Would you like to log in again?
Kimtech* A6 and A8 garments
10% off plus access to RightCycle* recycling programme
Kimtech* A6 and A8 garments
VALID UNTIL: 30/11/2017
Innovative, durable and breathable coveralls with enhanced comfort offering personal and process protection
Non sterile SMS-based coveralls from Kimberly-Clark Professional*. Two designs, with or without hood. Both designs have elastic cuffs and thumb loops, at least 94% bacterial filtration efficiency and are individually packed. The A6 design also has laminated SMS fabric on the hood, front panel, arms and legs and is EN 14126:2003 certified protection against infective agent penetration.
  • A6 - breathable, liquid protection, hooded coveralls
  • A8 - breathable, particle protection, unhooded coveralls
  • Certified PPE Cat III directive 89/686/EEC, type 5/6 and anti-static EN 1149-5:2008
  • Suitable for ISO 7/8 Grade C/D cleanrooms
  • Recycling programme is available for these Kimtech* garments (excludes Eastern Europe, Turkey and Italy), sign up to RightCycle® when you place your order
Colour Clothing size Pk Cat. No. Promo price Quantity
KIMTECH PURE* A6
White S 25 113-8614E
White M 25 113-8615E
White L 25 113-8616E
White XL 25 113-8617E
White XXL 25 113-8618E
White 3XL 25 113-8619E
White 4XL 25 113-8620E
KIMTECH PURE* A8
White S 25 113-8621E
White M 25 113-8622E
White L 25 113-8623E
White XL 25 113-8624E
White XXL 25 113-8625E
White 3XL 25 113-8626E
White 4XL 25 113-8627E