Would you like to log in again?
Non sterile cleanroom disposable overalls A6/A8
NEW!
Non sterile cleanroom disposable overalls A6/A8
Completing the well established A5 sterile cleanroom disposable overall range.
  • Non sterile - can be used in cleanroom ISO 7/8
  • SMS 45 gsm based
  • Certified Cat III type 5 and 6
  • Individually packed using compressed packaging
  • A6: Antistatic-treated to meet EN 1149-5 and EN 14126 infective agent protection
  • A6: Proprietary folding method to ensure that the outer surface of the overall is not touched when opening the packaging
Innovative, lightweight, durable and breathable, SMS offers comfort and chemical splash protection.
Size Pk Cat. No. Price Quantity
KIMTECH PURE A6 cleanroom overalls
Small 25 113-8614E
Medium 25 113-8615E
Large 25 113-8616E
X-Large 25 113-8617E
XX-Large 25 113-8618E
XXX-Large 25 113-8619E
XXXX-Large 25 113-8620E
KIMTECH PURE A8 cleanroom overalls
Small 25 113-8621E
Medium 25 113-8622E
Large 25 113-8623E
X-Large 25 113-8624E
XX-Large 25 113-8625E
XXX-Large 25 113-8626E
XXXX-Large 25 113-8627E