Would you like to log in again?
KAUTEX PVC wide neck containers
Special Offer!
KAUTEX PVC wide neck containers
VALID UNTIL: 31/10/2017
The transparent all-rounder
Ideal for packaging, sampling, storing and shipping pastes or powdered substances from -20 to +80 °C.
  • Easy to fill and clean: Wide neck up to 63 mm I.D.
  • Immediate display of the content: Available in clear or amber PVC
  • Space saver: Square shape design
  • Protect your samples: Leakproof PP screw caps, black with PE foam insert B24 (please order separately)
  • Suitable for food contact: Food conformity according regulation (EU) 10/2011
Wide neck containers, Series 310, PVC transparent, without screw cap
Capacity WxDxH Neck I-Ø Thread Pk Cat. No. Promo price Quantity
50 ml 38,5x38,5x59 mm 23 mm 32 mm 162 215-2185E
100 ml 46x46x71 mm 31 mm 40 mm 80 215-3116E
200 ml 59,5x59,5x87 mm 41 mm 50 mm 1 215-3906E
300 ml 67,5x67,5x94 mm 41 mm 50 mm 1 215-3907E
500 ml 80,5x80,5x108 mm 54 mm 65 mm 1 215-3908E
750 ml 91,5x91,5x120 mm 69 mm 80 mm 1 215-3910E
1000 ml 97x97x143 mm 69 mm 80 mm 1 215-3930E
2000 ml 122x122x187 mm 69 mm 80 mm 1 215-3931E
Wide neck containers, Series 310, PVC transparent, without screw cap
Capacity WxDxH Neck I-Ø Thread Pk Cat. No. Promo price Quantity
50 ml 38,5x38,5x59 mm 23 mm 32 mm 162 215-2186E
100 ml 46x46x71 mm 31 mm 40 mm 80 215-3118E
200 ml 59,5x59,5x87 mm 41 mm 50 mm 1 215-3916E
300 ml 67,5x67,5x94 mm 41 mm 50 mm 1 215-3917E
500 ml 80,5x80,5x108 mm 54 mm 65 mm 1 215-3918E
750 ml 91,5x91,5x120 mm 69 mm 80 mm 1 215-3920E
1000 ml 97x97x143 mm 69 mm 80 mm 1 215-3932E
2000 ml 122x122x187 mm 69 mm 80 mm 1 215-3933E
Closures for wide neck containers, Series 310, PVC
Capacity For bottles Pk Cat. No. Promo price Quantity
50 ml 32 mm 1 215-3921E
100 ml 40 mm 1 215-3922E
250 ml 50 mm 1 215-3923E
500/750 ml 65 mm 1 215-3924E
750/1000/1500/2000 ml 80 mm 1 215-3925E
2000 ml 80 mm 1 215-3926E