Would you like to log in again?
Product Ranges
Ultrasphere®, Partisil®, Partisphere®, Vydac®, Alltima®, Alltima® HP, Prevail™, Apollo, Allsep®, Apex™ and Genesis™ Columns
Hichrom is the proud manufacturers of the original Ultrasphere® range (formerly Beckman), the original Partisil® and Partisphere® ranges (formerly Whatman/GE Healthcare) and the original Vydac®, Alltima®, Alltima® HP, Prevail™, Apollo™, Allsep®, Apex™ and Genesis™ analytical HPLC column ranges (formerly Grace/Alltech).
Other Hichrom Columns
Hichrom C18 and C8 columns are based on ultra pure silica with advanced bonding technology and offer high performance for acidic, basic and neutral molecules.
Hichrom RPB is a high purity base deactivated silica offering unique selectivity due to its proprietary C8/C18 multi-alkyl bonding. It is specifically designed for the reversed-phase separation of basic compounds.
  • Hichrom C18 and C8 with 3.5 µm and 5 µm spherical silica particles
  • Hichrom RPB with 3.5 µm, 5 µm and 10 µm spherical silica particles
  • Excellent reproducibility
In addition to these phases, Hichrom can also manufacture columns from any commercially available silica and pack columns from customer supplied materials.
Distributed Column Ranges
Besides, our own manufactured products, Hichrom is an approved distributor and provides technical support for many other major HPLC column brands.
Please contact us for more information on these products: sales@hichrom.co.uk
Product Selection