Would you like to log in again?
ACE® HILIC – 3 reproducible proprietary HILIC selectivities unveiled
FREE HILIC wallchart and method development guide!
ACE® HILIC – 3 reproducible proprietary HILIC selectivities unveiled
VALID UNTIL: 31/10/2017
NEW phases and method development kits achieve successful, reproducible HILIC separations
3 novel phases: HILIC-A (acidic character phase), HILIC-B (basic character phase) and HILIC-N (neutral character phase).
  • A simplified and systematic approach to HILIC method development
  • Compatible with all manufacturers’ of HPLC and UHPLC systems
  • Cost effective method development kits with 3 columns for the price of 1
  • Ultra-robust columns up to 1000 bar (15 000 psi)
  • 3 particle sizes (1,7; 3 and 5 micron) in a wide range of dimensions