Would you like to log in again?
2100Q portable turbidimeter
20% Off!
2100Q portable turbidimeter
VALID UNTIL: 31/12/2017
Easiest calibration and verification with accurate results every time
Hach 2100Q portable turbidimeters offer unsurpassed ease of use and accuracy.
Includes instrument, case assembly, 4x AA alkaline batteries, 6 sample cells, Stablcal™ ampoule kit, silicone oil, oiling cloth, manual.
  • Easy on-screen assisted calibration and verification
  • Simple data transfer
  • Accuracy for rapidly settling samples
  • Convenient data logging
  • Optical system for precision in the field
Instrument model options
Description Cat. No. Promo price Quantity
2100Q IS portable turbidimeter, battery powered, MR 0,01 to 1,000 NTU - compliant with ISO 7027 design criteria 705-0670E
2100Q portable turbidimeter, battery powered, MR 0,01 to 1,000 NTU - compliant with USEPA Method 180.1 design criteria 705-0669E
Optional accessories
USB+ power module, includes module, universal power supply, standard USB cable, special USB cable adapter and manual 705-0914E
Further accessories are available on request