Would you like to log in again?
SL1000 portable parallel analyser
SL1000 portable parallel analyser
VALID UNTIL: 31/12/2017
Water quality testing dramatically streamlined
Perform common water tests with less than half the manual steps.
SL1000 meter, carrying case, 1 instrument sample cup, 2 electrode sample cups, rechargeable battery, battery charger, hand strap, instrument manual & USB cable.
  • Less variability with automated processes
  • Less headache with a field kit that requires fewer bulky accessories
  • Faster testing by 25%
  • Chemkeys® technology
Instrument model options
Description Features Cat. No. Promo price Quantity
SL1000 portable parallel analyser w/o Chemkeys® - create your own tailor-made system 705-0941E
SL1000 portable parallel analyser - starter kit 'Disinfection' SL1000 meter, carrying case, 1 instrument sample cup, 2 electrode sample cups, rechargeable battery, battery charger, hand strap, instrument manual and USB cable plus 1 pH electrode, 1 conductivity electrode and 1 box each of Free Chlorine, Total Chlorine, Monochloramine, Nitrite and Free Ammonia Chemkeys. 705-0942E
Reagents
Group I - Chemkey reagents for disinfection control
Description Range Pk Cat. No. Promo price Quantity
Free Chlorine* Chemkey® reagents 0,04 - 4,60 mg/l 25 705-0943E
Total Chlorine* Chemkey® reagents 0,04 - 4,60 mg/l 25 705-0945E
Ammonia and monochloramine Chemkey® reagents 0,50 mg/l; 4 - 4,60 mg/l 50 705-0948E
Nitrite Chemkey® reagents (6 months shelf life) 0,005 - 0,600 mg/l 25 705-0949E
Group II - Chemkey reagents for water treatment
Description Range Pk Cat. No. Promo price Quantity
Copper Chemkey® reagents 0,06 - 5,75 mg/l 25 705-0953E
Dissolved iron Chemkey® reagents 0,05 - 3,00 mg/l 25 705-0954E
High range total alcalinity Chemkey® reagent 200 - 700 mg/l 25 705-0955E
Low range totalal calinity Chemkey® reagents 20 - 200 mg/l 25 705-0956E
Low range hardness Chemkey® reagents 3 - 20 mg/l 25 705-0958E
High range orthophosphate Chemkey® reagents 2,0 - 30,0 mg/l 25 705-0959E
Low range orthophosphate Chemkey® reagents 0,20 - 4,00 mg/l 25 705-0960E
'*Other pack sizes are available including 300 tests.
Optional accessories
Description Cat. No. Promo price Quantity
Car charger for SL1000 portable parallel analyser 705-0963E
IntelliCAL„LDO101 Luminescent Dissolved Oxygen(LDO) sensor,1 m cable - measuring range 0,1 - 20 ml/l 664-0206E
Chlorine verification Chemkey® reagents 705-0961E
Verification system Chemkey® (12 months shelf life) 705-0962E