Would you like to log in again?
illustra™ ExoProStar™ S: Enzymatic PCR and sequence reaction clean-up
illustra™ ExoProStar™ S: Enzymatic PCR and sequence reaction clean-up
VALID UNTIL: 31/10/2017
Purify PCR and sequencing set-up reactions quickly, efficiently and reliably
ExoProStar S contains illustra Shrimp Alkaline phosphatase and Exonuclease A, formulated to work together to remove unincorporated primers and nucleotides from amplification reactions in preparation for sequencing, cloning, genotyping or further DNA modification reactions.
  • Enzymes provided in two separate tubes, just two simple pipetting steps are needed to prepare the reaction
  • Fast 15 minute protocol
  • Scalable for different reaction sizes
  • No loss of PCR product
  • Easy to automate
Description Pk Cat. No. Price Quantity
illustra ExoProStar S PCR and Sequence Reaction Clean Up Kit 100 R US79010E
illustra ExoProStar S PCR and Sequence Reaction Clean Up Kit 500 R US79050E
illustra ExoProStar S PCR and Sequence Reaction Clean Up Kit 2000 R US79200E