Would you like to log in again?
Safety Storage
Safety Storage
Cabinets for Flammable Liquids
The most important protection goal of a safety storage cabinet is to reliably shield flammables against a critical temperature increase (180K)  in the event of a fire over a defined period of time. Explosions can occur if the temperature rises by more than 180 K, because many flammable chemicals start to reach their auto ignitation point . Safety storage cabinets for flammable liquids are important to :         
  • Avoid explosions that accelerate the fire
  • Adequate time for personnel to escape
  • Adequate time for fire-fighting and rescue missions
Only Type 90 ( 90 minutes fire resistant ) cabinets in accordance with EN 14470-1 offer maximum safety.
Cabinets for Acids and Alkalis
Special cabinet for  safe storage of corrosive hazardous materials, especially acids and alkalis.Not suitable for the storage of flammable liquids.
Cabinets for Chemicals
Cabinet for safe and approved storage of chemicals, toxic materials and other substances that have to be kept under lock in workrooms.Not suitable for the storage of flammable liquids.
Gas Cylinder Cabinets
Maximum safety in the storage, provision and withdrawal of gas cylinders in indoor areas according to EN 14470-2
  • Type G90 fire resistant (90 minutes) gas cylinder cabinets
  • Type G30 fire resistant (30 minutes) gas cylinder cabinets