Would you like to log in again?
Laminar Air Flow Systems
Laminar Air Flow Systems Laminar flow cabinets are carefully designed to protect very sensitive products from external contamination or the user from contamination by the product - or both. This very clean working atmosphere is obtained by blowing air through HEPA filters. The airstream can be vertical or horizontal, depending on the application.
Product Selection