Would you like to log in again?
Global Web is currently experiencing issues, technical teams are working to correct the problem.
Kjeldahl Analysis
Kjeldahl Analysis
Kjeldahl nitrogen determinations are performed on a variety of food substances. The Kjeldahl method may be broken down into three main steps: digestion, distillation, and titration.
1. Digestion: is accomplished by boiling a homogeneous sample in concentrated sulphuric acid. The end result is an ammonium sulphate solution.
2. Distillation: excess base is added to the digestion product to convert the ammonium salt to ammonia.
3. Titration: the amount of nitrogen in a sample can be calculated from the quantified amount of ammonia in the receiving solution. The ammonia is captured either with a carefully measured excess of a standardised acid solution (back titration) or a boric acid solution (direct titration)