Would you like to log in again?
Eppendorf´s UVette®
Eppendorf´s UVette®
Eppendorf´s UVette® is a fully UV-transparent disposable cuvette.
The individually packaged cuvettes are certified DNase, RNase and protein-free and therefore the optimal choice for your valuable sample. The starter set contains a universal adapter to enable the use of the UVette in other instruments other than the Eppendorf BioPhotometer plus.
  • The UVette combines two optical pathlengths in one cuvette. This feature allows the UVette to cover measurements across a broad range of concentrations in a flexible manner
  • The funnel-shaped design of the UVette base prevents the capillary effect, thus ensuring that even the minimum volume of 50µL constantly remains in the centre of the measuring area
  • With the supplied universal adapter, the UVette fits optimally into nearly all spectrophotometers. The adapter aligns the UVette to the height of the photometer’s light beam and fulfils a diaphragm function for optimal light distribution
Technical specifications
Volume range, µL 50 – 2,000
Wavelength range, mm 220 – 1,600
Optical pathlengths, mm 10 and 2
External dimensions (WxDxH), mm 12.5 x 12.5 x 36
Light centre height, mm 8.5