Would you like to log in again?
FREE Multipette® M4 Offer
VWR Exclusive!
FREE Multipette® M4 Offer
VALID UNTIL: 30/04/2018
Order any 7 packs of Combitips advanced® and receive a free multipette® M4!
  • Ergonomic design with unique funnel shape prevents damage to gloves and ensures comfortable handling
  • Individually colour coded for quick identification
  • Elongated tips (for 2.5; 5 and 10 ml) allow complete emptying of all common tubes preventing sample loss
The Eppendorf Combitips advanced® have been completely redesigned and optimised to meet all the needs of any modern laboratory. Functioning according to the positive displacement principle, they always dispense the correct volume independently of the density of the liquid and its flow characteristics. Working with dangerous materials also becomes safer due to the hermetically sealed piston preventing aerosol contamination.
Delivery information: Please allow up to 10 days for delivery of your free Multipette® M4.