Would you like to log in again?
Turbidity
Turbidity
A simple and undeniable indicator of water quality change
The measurement of turbidity is a key test of water quality: drinking water – waste water monitoring – industrial waters and more…
Reliable turbidity monitoring is an essential component of drinking water production, and public health and safety organizations around the world recognize the importance of measuring this parameter.
A sudden change in turbidity may indicate an additional pollution source (biological, organic or inorganic) or may signal a problem in the water treatment process.
Are you having questions on the topic Turbidity or are you a beginner and need to know more?  Click below to download the "Theory of turbidity" booklet.
In practical applications significant problems can introduce interference and errors that reduce the accuracy of any instrument. To ensure the instrument is operating properly and providing the most accurate answers possible, it is important to verify its calibration.  Click below to download the "Technical information on standards"  booklet.
- Product Details
The Benchtop Turbidimeter TL23 series, Hach
Trusted measurement for wide range of turbidity applications
A turbidimeter that simplifies testing in the most demanding waste water and industrial applications
  • Easy to use intuitive full color touch screen
  • Easy data management and export capabilities
  • Easy to be right
Delivered with sample cells, standards, accessories
- Product Details
The new portable VWR TIR200
For fast, precise, easy On-Site testing
From drinking water to waste water, to be used for both colored and colorless liquids.  
Conforms to EN ISO 7027.
- Product Details
WTW, IDS turbidity sensors – digital & wireless ready
VisoTurb® 900-P: Digital IDS turbidity measurement with a practical turbidity sensor
  • Convenient turbidity sensor to measure turbidity in surface waters and process
  • Simple 2 or 3 point calibration
  • Suitable for WTW MultiLine® IDS multi-parameter measurements on site
Full range of turbidity meters
Full range of turbidity standards