Would you like to log in again?
Thermoreactors
Thermoreactors
Rapid digestion for COD and TOC.
3 models:
  • TR 320: basic model for 12 cuvettes and 7 pre-stored temperature programmes
  • TR 420: accepts 24 cuvettes and offers an additional 8 programmes defined by the user
  • TR 620: accepts 2 x 12 cuvettes in 2 temperature zones with 7 pre-stored and 8 user-defined programmes