Would you like to log in again?
Suspended Material
Suspended Material VWR offers a variety of products for you to measure water quality from the point of view of suspended materials: 
  • Rapid method with moisture analysers
  • Centrifugation or filtration methods in compliance with European norms
  • The method for removing volatile suspended solids from the mainly inorganic substances or fixed solids involves the use of both a furnace and an analytical balance.