Would you like to log in again?
Solutions For TOC Analysis
Solutions For TOC Analysis
Total Organic Carbon (TOC) Analysis Solutions for Detecting Organic Contamination in Water
Dissolved organic carbon is oxidised by sodium persulphate reagent and CO2 produced is measured by spectrophotometry. Many other reagents in solution are used: acids, bases ...depending on type of water analysed. The most common reagents can be supplied by VWR, for example:
  • Sodium hydroxide 1.2 N solution
  • Sulphuric acid 1.8 N solution
  • Sodium persulphate solutions, at various concentrations
  • Phosphoric acid 10% solution
  • Phosphoric acid 25% solution
Many other solutions are possible - please enquire.
Quick Access to Product Information and Ordering