Would you like to log in again?
Moisture Analysers
Moisture Analysers
Routine moisture analysers with IR and halogen heater type.
  • Optimum performance: repetitive results with readability up to 0.005 g, 0.05%
  • Simple to operate: Easy to set up, easy-to-use and inexpensive to maintain
  • Easy cleaning and modern robust design with great durability
  • 2 models: MB 23 uses an infrared coil (no glass) and MB 25 uses a halogen lamp