Would you like to log in again?
BOD Photometric Method
BOD Photometric Method
The BOD test is a photometric cell test from Merck
  • Calibrated and standard compliant analysis reagents for security
  • Bar code system enables rapid and simple operation
  • Comprehensive documentation of the quality control for every test
  • 5 days incubation still required but the analysis has fewer handling steps than other methods
Quick Access to Product Information and Ordering