Would you like to log in again?
Dilution BOD
Dilution BOD
BOD measurement official method according to EN 1899-1 and DI EN 1899-2, official EPA method.
  • Simple sample management
  • Automatic BOD calculation
  • EPA approved
Self-stirring oxygen sensor StirrOx® G for oxygen determination in Karlsruhe bottles allows:
  • Constant flow for high reproductibility
  • No zero point calibration necessary
  • Immediately ready for measuring