Would you like to log in again?
InVue™ CR288 concentration monitor - process monitoring for in-line liquid chemical applications
InVue™ CR288 concentration monitor - process monitoring for in-line liquid chemical applications
InVue™ CR288 concentration monitor offers real time information for point-of-use chemical/mixing blending, spiking and dilution without process intrusion or interruption.
  • Cost-effective and high performance concentration monitor enables greater process efficiency for BEOL, FEOL and subfab delivery chemical systems
  • Software integrates data collection, analysis, and field calibration, along with convenient, scalable data interface and acquisition options
  • LCD digital display unit (DDU) supports RS232 communication interface
  • Concentration monitor comes standard with fine thread Flaretek®, Super 300 Type Pillar® and PrimeLock® end connections
  • USP Class VI certified allows for safe integration into biological and pharmaceutical processes for in-line concentration monitoring
The CR288 uses an innovative refractive index technology packed in a compact and flexible configuration. The measurement is based on the refraction of light in the process fluid, resulting in an accurate, safe and repeatable means of measuring liquid concentrations.