Would you like to log in again?
EMSURE® salts for analysis with improved flowability – for pharmacopoeia analysis
NEW !
EMSURE® salts for analysis with improved flowability
Due to their natural hygroscopicity, many salts tend to cake. Merck has managed to improve the flowability of numerous EMSURE® salts for analysis simply by optimizing production processes in a decisive way. Since the dosage can be controlled more easily, these salts boost the reliability of analyses in the wet chemical lab substantially.
Merck EMSURE® salts for analysis with improved flowability
  • 1.06404 Sodium chloride for analysis EMSURE®, ACS, ISO, Reag. Ph Eur
  • 1.02382 Calcium chloride dihydrate for analysis EMSURE®, ACS, Reag. Ph Eur
  • 1.02790 Copper(II) sulfate pentahydrate for analysis EMSURE®, ACS, ISO, Reag. Ph Eur
  • 1.05833 Magnesium chloride hexahydrate for analysis EMSURE®, ACS, ISO, Reag. Ph Eur
  • 1.03965 Iron(II) sulfate heptahydrate for analysis EMSURE®, ACS, ISO, Reag. Ph Eur
  • 1.08883 Zinc sulfate heptahydrate for analysis EMSURE®, ACS, ISO, Reag. Ph Eur
  • 1.04936 Potassium chloride for analysis EMSURE®
Merck EMSURE® salts for analysis: The best specifications in the world
Merck EMSURE® salts for analysis are far ahead of the crowd with their high level of purity, often even exceeding international standards like ACS or ISO.
They not only fulfill the requirements of reagents for pharmacopoeia analysis, but also those of the American and European pharmacopoeia organizations. That’s why Merck’s EMSURE® salts for analysis also meet the high demands of customers who carry out quality assurance in the pharmaceutical industry. These reagents are designated as ACS and Reag. Ph Eur.
Merck EMSURE® salts for analysis: Certified quality management
Merck has DIN EN ISO 9001:2008 certification. Merck documents all production and analysis processes comprehensively. Each analysis certificate also indicates the best before date of the reagent. A high level of consistency across batches increases the reproducibility of analysis results.
If Merck EMSURE® salts for analysis should ever cake despite all this, gently shaking the packaging will restore the flowability of the salts.
Please wait while loading...
Description Pack Cat.No. Price Qty.
SODIUM CHLORIDE GR FOR ANALYSIS ACS, ISO 1 * 1 kg 1.06404.1000P
CALCIUM CHLORIDE-DIHYDRATE CRYST. GR 1 * 1 kg 1.02382.1000P
COPPER(II) SULPHATE 5-HYDRATE GR AC 1 * 1 kg 1.02790.1000P
IRON(II) SULFATE HEPTAHYDRATE GR ACS 1 * 1 kg 1.03965.1000P
POTASSIUM CHLORIDE GR 1 * 1 kg 1.04936.1000P
MAGNESIUM CHLORIDE HEXAHYDRATE GR ACS 1 * 1 kg 1.05833.1000P
ZINC SULPHATE-HEPTAHYDRATE GR ACS, ISO 1 * 1 kg 1.08883.1000P