Would you like to log in again?
Chemical protective garments: Factors to consider to make the right choice!
Chemical protective garments: Factors to consider to make the right choice!
Do you know what needs to be taken into account when selecting protective clothing?
  • Identify the hazard
  • Define the minimum levels of protection needed
  • Assess toxity of the hazardous substance
  • Define performance requirements of fabric and seams
Select PPE that is ergonomically designed, appropriate for the working conditions, has built-in comfort features and is made from wearer friendly materials. Read more about choosing the right PPE in the attached article!
Size Pk Cat. No. Price Quantity
Tyvek® 800J CHA5 coverall
Small 20 113-9716E
Medium 20 113-9717E
Large 20 113-9718E
X-Large 20 113-9719E
XX-Large 20 113-9720E
XXX-Large 20 113-9721E