Would you like to log in again?
Oxivir Plus - powerful detergent and broad spectrum disinfectant
Oxivir Plus - powerful detergent and broad spectrum disinfectant
Single step cleaning and disinfection, with no compromise on efficacy, thanks to the AHP (Accelerated Hydrogen Peroxide) technology.
  • Efficient descaling detergent, streak-free and scale retardant
  • Broad spectrum disinfectant with fast action on bacteria, fungi, yeast and viruses
  • Environmentally-friendly: Hydrogen peroxide naturally degrades into water and oxygen
  • Safe for the staff: Ready to use solution is non hazardous and has low VOCs
  • Compatible for use on water- and acid-resistant surfaces, as well as on fabrics
  • Available in a ready to use 750 ml spray bottle or as a concentrate in a 5 l canister
Oxivir Plus is a dual action product, saving time and money by combining excellent performance in cleaning and disinfection. It is effective on a broad range of pathogens, such as influenza, Norovirus, Hepatitis C, MRSA, etc. Oxivir Plus is particularly well suited for the routine decontamination of work surfaces and fabrics with a high infection risk.
Please note that these products are not available in all countries.