Would you like to log in again?
Corning® hybrigro SF™ Medium
Corning® hybrigro SF™ Medium
Increase hybridoma yields and production efficiency without using costly serum!
  • Superior growth and antibody production over serum supplemented media
  • Formulated with glutagro to provide a stable, ready to use formulation. No glutamine supplementation required
  • Contains Polaxamer 188 to provide shear force protection during agitated suspension culture
  • Formulated without phenol red
  • Contains low concentrations of highly purified non-animal origin recombinant proteins added for enhanced cell growth and productivity
  • Defined: There are no undefined hydrolysates, ultra filtered lysates, or peptides in the formulation
Corning hybrigro SF is a complete, animal component-free, defined medium specifically developed for serum-free growth and antibody production with a variety of hybridoma cell lines. 
Description Pack Cat. No. Price Quantity
Hybrigro SF Medium with Corning Glutagro™ supplement 500 ml bottles MDTH40-215-CVR