Would you like to log in again?
NeutraKlean - new sterile ready to use detergent from Contec
NeutraKlean - new sterile ready to use detergent from Contec
Keep your mind at ease and your A & B cleanrooms clean with this new sterile detergent
  • Gentle on surfaces: Neutral pH, low residue formulation
  • Gentle on people and the environment: No CLP hazard classification
  • Easy to use: Easy tear bags, no further dilution required, available in 1 l spray bottle or 5 l canister
  • Formulated with purified water (Ph. Eur.)
  • Validated gamma irradiation, minimum 25 kGy
  • Shelf life of 2 years when unopened, with an in-use shelf life of 6 months for the 1 l bag-in-bottle
NeutraKlean is designed for the removal of light soiling and disinfectant residues commonly found in cleanrooms. It also helps prevent the formation of biofilm. Fully documented and validated, NeutraKlean is suitable for use in grade A & B cleanrooms. Being ready and easy to use, it simplifies your SOPs and improves reproducibility.
Description Pk Cat. No. Price Quantity
NeutraKlean sterile trigger spray 6x 1 l 141-1305E
NeutraKlean sterile canister 2x 5 l 141-1306E