Would you like to log in again?
Consumable products for mini environment cleaning
Consumable products for mini environment cleaning
VALID UNTIL: 31/10/2017
Solutions to facilitate the cleaning and disinfection of isolators, glove boxes, laminar flow hoods, biosafety cabinets and RABS.
  • The EasyReach cleaning tool, with 100% quilted polyester pads, is designed to only yield a low level of particulates
  • EasyReach pads are available pre-saturated with sterile 70% IPA, so no spray bottles need to be passed into the controlled environment, and exactly the same amount of fluid is used by each operator
  • The VertiKlean MAX mop head frame can be used with a standard microfibre or sealed edge VertiKlean MAX mop head
  • This system can then be fitted with a choice of pre-saturated or dry wipes that can be saturated at the point of use
  • Both systems can be used with the autoclavable, short QuickConnect handle, which is only 400 mm long, so easily passes into a transfer hatch. Alternatively, a telescopic 400 mm handle, which can extend to 760 mm, is also available
The cleaning and disinfection of isolators, restricted access barrier systems (RABS) and laminar airflow cabinets can often be a difficult task. They are designed with operator or product safety in mind and so are not always easy to decontaminate. Contec offers a choice of cleaning tools which make cleaning and disinfection easier. 
Description Type Specification Size Pk Cat. No. Promo price Quantity
EasyReach mop head frame 2685 Autoclavable 19x6 cm 1 129-0572E
EasyReach cleaning pads MEQT0001 Dry, 100% knitted polyester 19x7 cm 4 heads x 15 bags 129-0574E
EasyReach cleaning pads MEQT0002 Dry, sterile, 100% knitted polyester 19x7 cm 4 heads x 15 bags 129-0575E
EasyReach cleaning pads PSME0001 Pre-saturated with 70% IPA, sterile, 100% knitted polyester 19x7 cm 8 heads x 8 pouches 129-0576E
VKMAX mop wipe frame 2651K Regular 19 cm 1 129-1194E
VertiKlean Max mop head VKMX0125 Standard 21x11x2 cm 4 mops x 12 bags 141-0989E
VertiKlean Max mop head VKMX0175 Standard, sterile 21x11x2 cm 1 mop x 48 bags 141-0990E
VertiKlean Max mop head VKSE0100 Sealed edge 24x13x1 cm 4 mops x 10 bags 129-1195E
VertiKlean Max mop head VKSE0150 Sealed edge, sterile 24x13x1 cm 1 mop x 40 bags 129-1196E
QuickConnect handle 2686 One piece, stainless steel 40 cm 1 129-0573E
QuickConnect handle 2642 Telescopic, anodised aluminium 40 - 75 cm 1 129-0202E
The VKMAX mop wipe frame, in conjunction with a sealed edge mop head, can be used with any 300x300 mm or larger wipe. Using a VertiKlean Max mop head, you can use any 230x280 mm or 300x300 mm wipe. Examples of suitable wipes are:
Sterile pre-saturated alcohol wipes, low endotoxin
Description Type Specification Size Pk Cat. No. Promo price Quantity
PROSAT wipes, meltblown polypropylene with 70% IPA PS-911LE Sterile 230x280 mm 30 wipes x 36 pouches 115-2439E
PROSAT wipes, Polynit Heatseal with 70% IPA PSWE0002 Sterile 300x300 mm 30 wipes x 15 pouches 115-1440E
PROSAT wipes, Polynit Heatseal with 70% IPA PSWE0003 Sterile 300x300 mm 10 wipes x 30 pouches 115-2440E
Sterile pre-saturated alcohol wipes
PROSAT wipes, meltblown polypropylene with 70% IPA PS-911EB Sterile 230x280 mm 30 wipes x 48 pouches 115-0487E
PROSAT wipes, meltblown polypropylene with 70% DE PS-911EB-ETOH Sterile 230x280 mm 30 wipes x 48 pouches 115-0546E
PROSAT wipes Amplitude Sigma with 70% IPA PSC20001 Sterile 230x280 mm 24 wipes x 48 pouches 115-0781E
PROSAT wipes Polynit Heatseal with 70% IPA PSPS0091 Sterile 300x300 mm 20 wipes x 20 pouches 115-2447E
Sterile dry wipes, low endotoxin
Amplitude Kappa LE wipes NWPZ0002 Sterile 300x300 mm 25 wipes x 40 pouches 115-1864E
Polynit Heatseal LE wipes LWLE0002 Sterile 300x300 mm 10 x 36 pouches 115-2448E
Sterile dry wipes
Polynit Heatseal wipes LWPS0007 Sterile 300x300 mm 25 wipes x 40 pouches 115-1862E
Quiltec I wipes LWQT0031 Sterile 300x300 mm 10 wipes x 50 pouches 115-2449E