Would you like to log in again?
Consumable products for endotoxin control
Consumable products for endotoxin control
VWR solutions to reduce bioburden and control endotoxin levels in your manufacturing process
  • Sterile alcohols and disinfectants (availability may vary per country so contact your local VWR customer service for more information)
  • Amplitude Kappa Sterile LE, dry non woven wipes, cost effective, double bagged in packs of 25 pieces, with an endotoxin limit of 20 EU per wipe
  • Polynit Heatseal Sterile LE, dry polyester wipes, with sealed edges, half folded and triple bagged in packs of 10 pieces, with an endotoxin limit of 1 EU per wipe
  • PROSAT® Sterile PS-911 LE, polypropylene wipes pre-saturated with 70% pharma grade IPA and 30% deionised water, double bagged in reclosable pouches of 30 pieces, with an endotoxin limit of 20 EU per wipe
  • PROSAT® Sterile Polynit Heatseal LE, sealed edge polyester wipes pre-saturated with 70% pharma-grade IPA and 30% Water for Injection (USP), triple bagged in reclosable pouches of 10 pieces, with an endotoxin limit of 1 EU per wipe
Endotoxins are difficult to remove from products once present. It is far better to keep finished products and components relatively free from endotoxins, rather than have to remove them afterwards. With Contec we offer a complete range of low endotoxin, contamination control products.
Description Material Dry/pre-saturated Packed Pk Cat. No. Price Quantity
Amplitude Kappa Sterile LE wipes; 23x23 cm, <20 EU/wipe Non woven polyester/lyocell blend Dry Double bagged, 40x25 wipes 1000 115-1863E
Amplitude Kappa Sterile LE wipes; 30x30 cm, <20 EU/wipe Non woven polyester/lyocell blend Dry Double bagged, 40x25 wipes 1000 115-1864E
Polynit Heatseal Sterile LE wipes; 23x23 cm, <1 EU/wipe Knitted polyester with sealed edges Dry Triple bagged, 50x10 wipes 500 115-2438E
PROSAT® Sterile PS-911 LE wipes; 23x28 cm, <20 EU/wipe Non woven polypropylene 70% pharma grade IPA 30% deionised water Double bagged, 36x30 wipes 1080 115-2439
PROSAT® Sterile Polynit Heatseal LE wipes; 23x23 cm, <1 EU/wipe Knitted polyester with sealed edges 70% pharma grade IPA 30% water for injection Triple bagged, 55x10 wipes 550 115-1424E
PROSAT® Sterile Polynit Heatseal LE wipes; 30x30 cm, <1 EU/wipe Knitted polyester with sealed edges 70% pharma grade IPA 30% water for injection Triple bagged, 30x10 wipes 300 115-2440