Would you like to log in again?
Contec range of cleanroom wipes, mops, detergents and disinfectants
More than meets the eye!
Contec range of cleanroom wipes, mops, detergents and disinfectants
Mission-critical cleaning and contamination control
  • Dry and pre-saturated, knitted and non woven wipes specifically designed for pharmaceutical applications
  • Extensive mopping range - newly added curtain cleaner and sealed edge mops
  • Completed range of alcohols, disinfectants and detergents
From patented Anticon wipes with particle attraction technology, or market leading pre-saturated wipes in a variety of substrates, Contec have a cleanroom wipe to suit every budget, application and facility. Plus a newly completed range of alcohols, disinfectants and detergents. Contec have also launched three new innovative mopping products, adding a curtain cleaner and sealed edge mops to their extensive mopping range.
Description Size Material Sterile/autoclavable Pk Cat. No. Price Quantity
EasySat bucketless floor mop system
Mop head EasySat, smooth 55 cm PU foam & smooth polyester Non sterile 32 129-0213
Mop head EasySat, smooth 55 cm PU foam & smooth polyester Sterile 28 129-0214
Mop head EasySat, textured 55 cm PU foam & textured polyester Non sterile 32 129-0215
Mop head EasySat, textured 55 cm PU foam & textured polyester Sterile 28 129-0338
Bucketless Floor Mop System EasySat, with 1,5 l tank, autoclavable Autoclavable 1 141-0100
VertiKlean Max floor and wall mop system
Mop head VertiKlean Max, regular 21 cm PU foam & microfibre Non sterile 48 141-0989
Mop head VertiKlean Max, regular 21 cm PU foam & microfibre Sterile 48 141-0990
Mop head VertiKlean Max, medium 32 cm PU foam & microfibre Non sterile 28 141-0991
Mop head VertiKlean Max, medium 32 cm PU foam & microfibre Sterile 28 141-0992
Mop head VertiKlean Max, large 38 cm PU foam & microfibre Non sterile 24 141-0993
Mop head VertiKlean Max, large 38 cm PU foam & microfibre Sterile 24 141-0995
Frame for mop head VertiKlean Max regular Stainless steel Autoclavable 1 129-0207
Frame for mop head VertiKlean Max medium & large Stainless steel Autoclavable 1 129-0208
Handle QuickConnect 40 - 75 cm Aluminum 1 129-0202
Handle QuickConnect 130 - 230 cm Aluminum 1 129-0203
Handle QuickConnect 40 - 75 cm Stainless steel Autoclavable 1 129-0204
Handle QuickConnect 130 - 230 cm Stainless steel Autoclavable 1 129-0205
Cart double bucket, 1 red 20 l + 1 grey 20 l, frame with castors Polypropylene & stainless steel Autoclavable 1 141-0969
Wringer flat for VertiKlean Max mop, requires 141-0966 Stainless steel Autoclavable 1 141-0967
Frame for wringer 141-0967, fits on 20 l bucket Stainless steel Autoclavable 1 141-0966
Pre-saturated wipers, sterile and non sterile
Wiper SatWipes C3, 100% IPA, snap-close canister 15x23 cm Non woven polyester Non sterile 1200 115-0379
Wiper ProSat 911, 70% IPA/30% DIW, resealable pouch 23x28 cm Meltblown polypropylene Non sterile 30 115-0270
Wiper ProSat 911, 70% IPA/30% DIW, resealable pouch 23x28 cm Meltblown polypropylene Sterile 1440 115-0487
Wiper ProSat 911, 70% Denat. Ethanol/30% DIW, resealable pouch 23x28 cm Meltblown polypropylene Non sterile 1440 115-0546
Wiper ProSat 911, 70% Denat. Ethanol/30% DIW, resealable pouch 23x28 cm Meltblown polypropylene Sterile 30 115-1422
Wiper ProSat Sigma, 70% IPA/30% DIW, resealable pouch 23x28 cm Polyester/cellulose Non sterile 1200 115-1423
Wiper ProSat Sigma, 70% IPA/30% DIW, resealable pouch 23x28 cm Polyester/cellulose Sterile 1152 115-0781
Wiper ProSat PNHS, 70% IPA/30% DIW, resealable pouch 23x23 cm Knitted polyester sealed edges Non sterile 30 115-1865
Wiper ProSat PNHS, 70% IPA / 30% WFI, resealable pouch, low endotoxins 23x23 cm Knitted polyester sealed edges Sterile 550 115-1424
Dry wipers, sterile and non sterile
Wiper Delta 10x10 cm Non woven polyester Non sterile 1200 115-1851
Wiper Delta 23x23 cm Non woven polyester Non sterile 3600 115-1850
Wiper Zeta 20x23cm Polypropylene/cellulose Non sterile 150 115-0610
Wiper Zeta 28x30 cm Polypropylene/cellulose Non sterile 75 115-0611
Wiper Epsilon, blue 23x23 cm Polyester/cellulose Non sterile 300 115-1853
Wiper Sigma 23x23 cm Polyester/cellulose Non sterile 300 115-0573
Wiper Sigma 30x30 cm Polyester/cellulose Non sterile 150 115-0574
Wiper SterileSorb 23x23 cm Polyester/cellulose Sterile 1500 115-0498
Wiper SterileSorb 30x30cm Polyester/cellulose Sterile 1050 115-1856
Wiper PolyNit HeatSeal 23x23 cm Knitted polyester sealed edges Non sterile 150 115-0268
Wiper PolyNit HeatSeal 30x30 cm Knitted polyester sealed edges Non sterile 75 115-0375
Wiper PolyNit HeatSeal 30x30 cm Knitted polyester sealed edges Sterile 1000 115-1862
Wiper Quiltec1, 2-ply 23x23 cm Knitted polyester sealed edges Non sterile 100 115-0378