Would you like to log in again?
Accredited pipette calibration and repair service
Accredited pipette calibration and repair service
Fast, reliable, competent
VWR‘s manufacturer independent pipette calibration service provides traceable results to the highest standards to ensure your instruments are compliant. 
Your benefits:
  • Proven and verified quality
  • Detailed process documentation
  • All manufacturers
  • All liquid handling instruments including glassware
  • All type of examinations including customer specifications
  • Calibration with certification up to and including a UKAS certificate
  • Creation of a as-found and as-left certificate
  • Use of original manufacturer spare parts
Request for Quote
Based on 20 years of experience for calibrating, repairing and servicing pipettes, we have created Pipette Insurance Plans. These plans are a complete package tailored to meet your needs:
VWR Light – economic calibration and adjustment
If for your pipettes are calibrated several times a year, maintenance might not be needed with every calibration. This PIP plan answers to this need and can be easily combined with VWR Care, VWR Eco or VWR Pro.
VWR Eco – preventative maintenance, economic calibration and adjustment
When you do not need to compy with an ISO standard, VWR Eco is an economic solution for your pipettes. With an as-left calibration of 2*4 measurements and an extensive preventative maintenance, we guarantee your pipette is in perfect order at an economical cost.
VWR Pro – preventative maintenance, economic calibration and adjustment according to ISO8655
As with the VWR Eco, we conduct extensive preventative maintenance to ensure your pipettes are always in perfect condition. Additionally we conduct an as-left calibration of 3*10 measurements according ISO 8655.
VWR Compliant – GLP/GMP total insurance plan according to ISO8655
Before we carry out the preventative maintenance we will execute an as-found calibration with 3*10 measurements. The as-found calibration is followed by preventative maintenanc e and an as-left calibration with 3*10 measurements. You will receive 2 calibration certificates, one of the as-found calibration and one of the as-left calibration.
VWR Care – ensuring your pipette is in top condition
Calibration is a standard in quality assurance, but what about preventative maintenance? VWR Care is for those customers who calibrate the pipette regular, themselves, but who do not do preventative maintenance. In this package we will carry out the preventative maintenance without doing a calibration. With this package you keep your pipette in top condition but it does not assure yet that accuracy and repeatibility is within specification.
Download order form
All our technicians are trained and certified by the manufacturers and our service center in the Netherlands. Due to our ISO 17025 certification we can offer on-site calibration or, alterantively at our customer's site. VWR is the only company certified on all pipette manufacturers to the standards as stipulated by FDA, ISO and GMP/GLP requirements.