Would you like to log in again?
Zero Air Generators
Zero Air Generators Zero air is commonly used as an oxidant for GC-FID or as a support gas for Total Hyrocarbon analysers. In both these examples it is important that the hydrocarbon level is minimised due to reduced maintenance (FID) or low baselines.
Zero Air Generators
  • Zero Air and Ultra Zero Air generators and stations
Zero Air Generators, VWR Collection Linea 10
Zero Air Generators
  • High quality hydrocarbon-free air for GC FID detectors
  • Requires an external compressed air source
Ultra Zero Air Generators
  • Supplies hydrocarbon-free air
  • Removes particles, moisture, CO2, CO, NOx, SO2 and O3 contaminants to less than 0.1 ppm
  • Requires an external compressed air source
Zero Air and Ultra Zero Air Stations
  • Contains an additional built in air compressor
Zero Air Generators, Parker Domnick Hunter
UHP Series
Parker Domnick Hunter zero air generators produce a continuous flow of clean dry air, with an ultra low residual hydrocarbon content of less than 0.05 ppm from an existing compressed air supply.
An interchangeable top panel allows for direct mounting to provide an all in one flame gas solution for GC-FID, FPD and NPD applications.
The generators can also be used to supply zero air to many additional applications. These include nebulisers and exhaust gas for LC/MS applications, zero gas and combustion gas for total hydrocarbon analysers and calibration/dilution gas for gas sensing equipment.
  • High performance catalyst
  • Oil-X Evolution pre- and after filtration
  • Digital temperature display
  • Minimal maintenance