Would you like to log in again?
Gas Chromatography
Gas Chromatography
Gas Chromatography employs partition between a gas phase and a functionalised immobilised liquid phase to achieve separation.  Owing to the length of the capillary columns, high resolution can be achieved.  
Gas Chromatography, capillary gas chromatography or just GC is widely used in the oil industry, food testing for pesticide analysis, pharmaceutical industry for residual solvent testing and many other applications.  It is mainly used for the analysis of volatile or semi volatile analytes.
Product pages