Would you like to log in again?
Instrument Systems and Modules
Instrument Systems and Modules
La Prep Sigma - The Sum of Possibilities
The new compact preparative chromatography system from VWR
  • modular system
  • many possible configurations
  • smallest foot print in the market
  • easy-to-use touch screen panel
  • ethernet connection
  • EZChrom software platform
  • pump heads with Titanium inlays for all sample contact components
  • double-piston pump active piston seal wash
  • electronic residual pulsation suppressor and multi-purpose valves
  • optional combination with autosampler and/or fraction collector
    manual or fully automated control
  For any enquiries, speak to your local VWR contact or email  chromatography@eu.vwr.com