Would you like to log in again?
Nitrogen Generators and LC-MS Nitrogen Generators, VWR Collection Linea 10
Nitrogen Generators and LC-MS Nitrogen Generators, VWR Collection Linea 10
The N2 Bora series generators are ideal for high purity laboratory nitrogen supply and chromatography or LCMS applications. Other typical applications are ICP, ELSD, PID or incubators.
  • Small units
  • PSA (Pressure Swing Adsorption) technology
  • N2 with a high purity of up to 99.999%.
  • Optionally with internal oil-free air compressor
N2 Sirocco
Series generators for high purity nitrogen at large flow rates:
  • DS-PSA (Double Stage PSA) technology
  • Most N2-Sirocco models include an internal oil-free compressor
  • N2 with a purity of up to 99.999%
Mistral-LCMS
  • Specially designed to maintain LC-MS systems
  • Up to 35 l/min nitrogen with a purity of 98.5%
  • PSA (Pressure Swing Adsorption) technology
  • Includes an internal compressor
N2-Mistral-0
  • Designed for LCMS applications
  • 98% purity
  • Based on hollow fibre membrane technology
  • Requires an external compressed air supply
  • Works pneumatically, no power supply needed
  • Completely silent
  • No moving parts, resulting in reliable and trouble-free operation with virtually no maintenance
N2 Sirocco large flow rate high purity of nitrogen generators:
  • DS-PSA (Double Stage PSA) technology
  • Most N2-Sirocco models include an internal oil free compressor
  • N2 with a purity of up to 99.999%
Mistral-LCMS:
  • Specially designed to compliment LC-MS systems
  • PSA (Pressure Swing Adsorption) technology
  • Including an internal compressor
  • Up to 35 L/min Nitrogen with a purity of 98.5%
N2-Mistral-0:
  • Designed for LCMS applications
  • Based on hollow fiber membrane technology
  • Requires external compressed air supply
  • Works pneumatically, no power supply is needed
  • Completely silent
  • No moving parts, resulting in reliable and trouble free operation with virtually no maintenance
Nitrogen Generators, Parker Domnick Hunter
Parker Domnick Hunter nitrogen generators are suitable for a wide range of laboratory applications, from purge gas for ICP-OES instruments to applications where purity is less critical such as ELSD, GC make-up gas, NMR sample spinning and Circular Dichroism.
All generators employ proven durability pressure swing adsorption (PSA) technology to deliver a  best in class life expectancy. The technology uses a carbon molecular sieve to remove oxygen, carbon dioxide and moisture from a compressed air supply to produce laboratory grade nitrogen.
An integrated oil-free compressor is optional, but all generators provide nitrogen at purities ranging from 98% to 99.999% (or better). Zero nitrogen and nitrogen and air generators are also available for more specific GC applications.
LC/MS Nitrogen Generators, Parker Domnick Hunter
Parker Domnick Hunter LC/MS nitrogen generators are specifically designed to meet the gas flow, purity and pressure requirements of the latest generation of LC/MS instruments.
Utilising the pressure swing adsorption technique, the generators provide a continuous stream of clean, dry nitrogen without the need for secondary purification and are fully compatible with all known APCI and ESI interfaces.
  • Models featuring integral oil-free compressors give increased security of supply, completely removing the reliance on external air supplies.
  • Suitable for nebulising, sheath and drying gas
  • Fully regenerative, proven durability with PSA technology
  • Phthalate-free components
  • Digital interface