Would you like to log in again?
Hydrogen Generators, VWR Collection Linea 10
Hydrogen Generators, VWR Collection Linea 10
In addition to their high safety (production of hydrogen from pure water only when required), the new VWR Collection LINEA10 hydrogen gas generators have been designed to meet highest quality and reliability standards. They employ the newest PEM membrane technology available for electrolytic production of pure hydrogen gas, including exclusive no maintenance auto-drying technology.
  • "No Maintenance" systems
  • Compact
  • Safe
  • Ergonomic and cost effective
  • Cascading option: combination and control of several hydrogen generators in series
  • Remote control option
Application fields:
H2 FID model series:
  • Fuel gas for GC-FID detectors
  • Reactor gas for other types of detectors in gas chromatography
H2 CARRIER model series:
  • Hydrogen as carrier gas in GC
  • Total hydrocarbon analysers
  • Sulphur analysers
  • Air-pollution monitoring systems
Hydrogen + Zero Air Station:
The H2+Air FID Station has been designed to feed GC-FID detectors with
  • Hydrocarbon free hydrogen and hydrocarbon free Zero Air at the same time
  • It has a small footprint (unit itself + external water tank)
  • Requires an external source of compressed air
H2 FID model series
  • Fuel gas for GC-FID detectors
  • Reactor gas for other types of detectors in gas chromatography
H2 CARRIER model series
  • Hydrogen as carrier gas for GC
  • Total hydrocarbon analysers
  • Sulphur analysers
  • Air pollution monitoring systems
Hydrogen & Zero Air Station
The H2 and Air FID station has been designed to feed GC-FID detectors with
  • Hydrocarbon-free hydrogen and hydrocarbon-free zero air at the same time
  • Small footprint (unit itself plus external water tank)
  • Requires an external source of compressed air
Hydrogen Generators, Parker Domnick Hunter
H-MD series:
Ultra high purity: 99.9999% H₂; 0-7 bar for GC carrier gas applications
H series:
High purity: 99.999 H₂; 0-7 bar for GC fuel gas applications
These generators are ideal for supplying fuel gas to all known combustion detectors used routinely in GC and THA, and can also be used to supply hydrogen for GC carrier gas applications and for ELCD reaction gas.
  • Automatic hydrogen leak detection
  • Patented seven stage explosion protection system
  • Continuous monitoring of "vital parameters"
  • Easy-to-use controller, high visibility/audibile alarms
  • Largest capacity water tank/optional auto water fill
  • Optional remote control and monitoring