Would you like to log in again?
Instrument Systems and Accessories
The innovative Perkin Elmer series of GC and GC-MS systems
Instrument Systems and Accessories
Clarus® 600 GC, Perkin Elmer
High performance, fast and reliable
  • Best in class oven enhances productivity: cooling from 450 to 50 °C in 100 seconds without cryogenics
  • Injector with PSS temperature control
  • Intuitive touch-screen interface and automatic steering
PPC = Programmable Pneumatic Control
Clarus 600 GC/MS, Perkin Elmer
  • Widest mass range of  1 to 1200 Da
  • 4 pump models, vacuumpumps, Turbo molecular and diffusion from 60 to 255 l/s
  • Quadrupole mass spectrometer, El+, Cl+, Cl-
  • Fast scan speed 12 500 amu/s, «Fast GC» compatible
  • SIFI™ simultaneous collection of selected ion and full ion scanning data
Clarus 500 GC, Perkin Elmer
The Clarus® 500 features a full colour, easy-to-use touch screen interface, so virtually no staff training is required
  • Flexible configurations with integrated mass spectrometry, headspace sampling or thermal desorption
  • Rugged, proven design provides reliable performance year after year
PPC = Programmable Pneumatic Control
TurboMatrix HS
Headspace autosamplers with or without analyte trap
HS 16 - HS 40 - HS110
  • 3 models with sampling capacities of 16, 40 and 110 samples
  • Manual or programmable pneumatic control
HS Trap 40 - HS Trap 110
  • Models with built-in analyte-trapping capability
TurboMatrix TD
Thermal desorbers for single tube or with an autosampler for 50 tubes
100 TD - 150 ATD - 300TD - 350 ATD - 650 ATD
  • Peltier cooling to 30 °C
  • Electronic gas regulation (150, 350 and 650 ATD) by integrated mass flowmeters
  • Automatic sample re-collection capability (TurboMatrix 650 only) enables repeat analysis
EZChrom Elite Software Chromatography Data System
 A uniform and scaleable software solution
  • Easy to operate
  • Instrument control
  • Data processing
  • Regulatory compliance
  • Network scalability
  • Dedicated driver for Perkin Elmer GC's
EZChrom Elite is one of the most popular chromatography data systems in the world. It complies with today’s demands in analytics - from a single workstation to a fully equipped and companywide client/server system. EZChrom Elite is a userfriendly, intuitive software developed for the current requirements of the pharmaceutical and chemical industries. It includes special tools for compliance with regulatory demands, e.g. FDA 21CFR11 and instrument and system validation.
EZChrom Elite Brochure