Would you like to log in again?
Chromatography Data Systems
Chromatography Data Systems Today nearly all users of HPLC and GC systems invest in a data system to control their instruments. Many laboratories have a mixture of instrumentation which ideally requires a software platform offering extensive control and data management over a wide range of instrument systems from different vendors. For this purpose VWR offers a range of professional Chromatography Data Systems (CDS) for Workstation and Client/Server installations in the regulated and non-regulated lab. Full control of VWR-Hitachi HPLC systems is provided as well.
Such an add-on package allows full control of the Hitachi LC’s within the different data systems. The Hitachi LaChrom Ultra and LaChrom Elite system runs along with instruments from other manufacturers in one software platform. This standardizes your operations with a single software interface to manage your data sets, instrument parameters, sequences, methods and reports.
The control for the Hitachi LaChrom Elite and Hitachi LaChromUltra instrument has been developed by Hitachi engineers and they have leveraged the software technology from the software manufacturer to implement the instrument control. This is especially important to protect the user from unwanted failures. It ensures that all instrument capabilities are included and full support is given by the Hitachi engineers. Needless to say that reverse-engineering technology wasn’t necessary to develop the instrument control. Only validated controls ensure a secure and reliable instrument control lamp usage and replacements, audit trails, automatic documentation of the instrument parameters, Maintenance parameters, etc.
OpenLAB CDS EZChrom Edition
A quantum leap in the evolution of EZChrom Elite
New scalable and compliant chromatography software platform with multi-instrument control
  • Scalable from workstations to fully featured client/server systems
  • Configurable compliance
  • Multi-instrument control
  • Intelligent reporting
  • Result Package mode
  • Many other new features
OpenLAB CDS EZChrom Edition is part of the new Agilent software suite OpenLAB for the analytical laboratory and enterprise. Its multi-instrument control capabilities, scalability and compliance are ideal to fulfil the requirements of today’s chromatography laboratories and to optimise their efficiency. Central administration functions have been arranged in the OpenLAB Shared Services/Control Panel module while keeping the existing user interface and workflows for the EZChrom Elite chromatography data system. Many exciting new functions are now available, from the Result Package mode and Result Review/Result Sequence mode to the new Intelligent Reporting mode. OpenLAB CDS EZChrom Edition can be easily complemented by further software modules belonging to the OpenLAB software suite. OpenLAB and EZChrom Elite are registered trademarks of Agilent Technologies, Inc.
Download Flyer
VWR-Hitachi LC Control for Waters Empower Software
The VWR-Hitachi LC’s run along with instruments from other manufacturers in the Empower software platform. This standardises your operations by using a single software interface to manage your data sets, instrument parameters, sequences, methods and reports.
VWR-Hitachi LC Control Software for Bruker Compass™/HyStar™ LC-MS Software
Compass™ is Bruker Daltonic´s unified software environment for mass spectrometric instrument control and data processing.
The Bruker Daltonics Compass™/HyStar™ software for LC/MS features plug-in based LC instrument control interfaces. This allows users to operate the following mass spectrometers in combination with the VWR-Hitachi LC systems:
  • amaZon series ion trap systems
  • micrOTOF™
  • micrOTOF-Q™
All systems are supported by the powerful Compass™ software which allows the user to easily handle all aspects of MS and LC system control and handle data with an intuitive and easy-to-use graphical user interface.