Would you like to log in again?
Cleanroom Bucket Systems
- Plastic -
Bucket Systems - Plastic
Plastic cleanroom cleaning equipment is available as single- or double-bucket systems. The bucket and squeezers are made from high quality, sturdy plastic (polypropylene). Buckets with litre scale, cart with steering rollers. Alternative models with flat squeezer are available on request.
We recommend bucket liners for easier transport of liquids and waste disposals out of controlled areas. They are gamma irradiated and single sterile packed.
Special accessories:
Bucket liners create an impermeable barrier between disinfectant and receptacle.