Would you like to log in again?
BRAND Ultra Low Retention Tips, BIO-CERT® – More hydrophobic than PTFE
BRAND Ultra Low Retention Tips, BIO-CERT® – More hydrophobic than PTFE
Extremely liquid repellent surface - produced through a special, optimized physicochemical process.
  • Ideal for biological samples that contain detergents such as Triton® X-100, SDS, Tween etc.
  • No additives that can be leached out! No siliconization of the surface!
  • High chemical resistance. Ideal for working with solvents.
  • The tips and boxes can be autoclaved at 121 °C (2 bar) without damaging the material properties.
  • BIO-CERT® quality certifies sterility and freedom of DNA, RNase, endotoxins and ATP.
The wetting of pipette tips when pipetting aqueous liquids containing detergents like SDS or Tween 80 plays a critical role in the life sciences. Based on photometric measurements, BRAND Ultra Low Retention tips were compared with low retention tips from various manufacturers. The results clearly show that the BRAND Ultra Low Retention tips exhibit up to 16 times less residues, although the pipetting steps were performed by a variety of different people with various levels of experience in pipetting.