Would you like to log in again?
BBP™12 label printer laboratory Identification
BBP™12 label printer laboratory Identification
Identify every sample with the BBP12 label printer!
Brady’s new, compact and affordable BBP12 label printer prints sharp and durable identification labels for vials, tubes, bottles, conicals, slides, straws, tissue cassettes and many more. Identify almost every sample with the BBP12 to avoid 'sticky situations'!
Discover 12 reasons why the BBP12 is a great choice!
A wide range of labels
The BBP12 is an entry level benchtop printer with an incredible amount of label options optimised for different environments. Able to print on label materials that resist liquid nitrogen, heat and chemicals, the standard BBP12 can print a great many lab labels, ranging from 10 to 112 mm wide to identify everything from microcentrifuges to large conicals. When coupled with the free label unwinder, the amount of labels is expanded even further. This enables the small BBP12 to print a durable identification label for almost any laboratory sample, in any environment.
Fits on almost every desk
The BBP12 is an extremely compact at only 26 cm long and 20 cm wide. It is designed to fit on almost every desk and occupies little space while opening up a lot of sample labelling capabilities.
Great value for money
The BBP12 is an ‘all-inclusive’ printer with extended standard features that offer a great many benefits. It is an excellent solution to avoid hidden upgrade costs. The printer’s benefits include:
  • 300 dpi quality printing
  • Easy calibration
  • Ethernet connectivity for use in networks
  • Connect any keyboard for stand-alone mode
Want to learn more? Discover all 12 reasons to choose the BBP12!