Would you like to log in again?
Drop. Read. Done. Little Genius!
Drop. Read. Done. Little Genius!
Introducing the BLItz System from Pall Life Sciences Protein Presence.
Quantitation. Kinetics. Assay Development.
The BLItz system provides a simple, rapid Dip and Read™ approach to protein analysis in an affordably priced personal assay system. Protein and antibody detection can be performed in a matter of seconds with high specificity and sensitivity, even in crude samples. The BLItz system enables real-time, label-free analysis of interactions on the surface of disposable fibre optic biosensors. Affinity, concentration and binding kinetics can be measured right at the bench in a 4 μL drop of sample.
BLItz measurements can be used to complement and in some cases entirely replace more labour-intensive methods to assess:
  • Binding pair optimization
  • Ligand binding studies
  • Investigation of process conditions
  • Quantitative and kinetics studies
A variety of disposable, off the shelf biosensors are available for BLItz, making it easy to run a wide range of assays on the same system.
Request your Free Demonstration of the Blitz system now – contact vwrbiomarke@eu.vwr.com