Would you like to log in again?
Discover the complete range of Biowest most used Cell Culture Media
Save 25%
Biowest Cell Culture Media formulations are manufactured according to original publications, standards set by the Tissue Culture Association and accepted formulations.
Formulations may vary from set standards by substituting hydrated, chlorinated and/or the salt forms of certain compounds.
These substitutions provide improvements in terms of product performance
DMEM - Dulbecco's Modified Eagle Medium
Dulbecco's Modified Eagle Medium (DMEM) is designed to preserve and maintain the growth of a broad spectrum of mammalian cell types.
Discover the wide range of Biowest Dulbecco's Modified Eagle Medium
  • DMEM High Glucose
  • DMEM Low Glucose
  • DMEM - Ham's F12
RPMI 1640 Medium
RPMI 1640 was developed by Moore and his co-workers at Roswell Park Memorial Institute in 1966 for the growth of human leukemia cells in monolayer or suspension cultures. RPMI Media is a "multipurpose" enriched media providing extensive applications for a broad spectrum of mammalian and hybridoma cells including:
  • Human myeloma
  • Mouse hybridoma
  • Human leukocytes
  • B and T lymphocytes
Discover the complete range of Biowest RPMI 1640
Description Cat. No. Promo price Quantity
DMEM - F12 w/ L-Glutamine w/ 15 mM Hepes - 500ml L0093-500P
DMEM - F12 w/ L-Glutamine w/ 25 mM Hepes - 500ml L0095-500P
DMEM - F12 w/ L-Glutamine w/o Sodium Bicarbonate w/ 15 mM Hepes - For 10L P0095-N10LP
DMEM - F12 w/ L-Glutamine w/o Sodium Bicarbonate w/ 15 mM Hepes - For 1L P0095-N1LP
DMEM - F12 w/ L-Glutamine w/o Sodium Bicarbonate w/ 15 mM Hepes - For 50L P0095-N50LP
DMEM - F12 w/ stable Glutamine w/ 15 mM Hepes - 500ml L0092-500P
DMEM - F12 w/o L- Glutamine w/o Hepes - 500ml L0090-500P
DMEM - F12 w/o L-Glutamine w/ 15 mM Hepes - 500ml L0094-500P
DMEM - F12 w/o L-Glutamine w/ 25 mM Hepes - 500ml L0096-500P
DMEM - F12 w/o L-Glutamine w/o Hepes w/o Glucose - 500ml L0091-500P
DMEM High Glucose w/ L-Glutamine w/o Sodium Bicarbonate w/o Sodium Pyruvate - For 10L P0103-N10LP
DMEM High Glucose w/ L-Glutamine w/o Sodium Bicarbonate w/o Sodium Pyruvate - For 1L P0103-N1LP
DMEM High Glucose w/ L-Glutamine w/o Sodium Bicarbonate w/o Sodium Pyruvate - For 50L P0103-N50LP
DMEM High Glucose w/ L-Glutamine w/o Sodium Bicarbonate w/o Sodium Pyruvate - For 5L P0103-N5LP
DMEM High Glucose w/ L-Glutamine w/ Sodium Pyruvate - 500ml L0104-500P
DMEM High Glucose w/ L-Glutamine w/o Sodium Bicarbonate w/ Sodium Pyruvate - For 10L P0102-N10LP
DMEM High Glucose w/ L-Glutamine w/o Sodium Bicarbonate w/ Sodium Pyruvate - For 1L P0102-N1LP
DMEM High Glucose w/ L-Glutamine w/o Sodium Bicarbonate w/ Sodium Pyruvate - For 50L P0102-N50LP
DMEM High Glucose w/ L-Glutamine w/o Sodium Bicarbonate w/ Sodium Pyruvate - For 5L P0102-N5LP
DMEM High Glucose w/ L-Glutamine w/o Sodium Pyruvate - 500ml L0102-500P
DMEM High Glucose w/ stable Glutamine w/ 25mM Hepes w/o Sodium Pyruvate - 500ml L0107-500P
DMEM High Glucose w/ stable Glutamine w/ Sodium Pyruvate - 500ml L0103-500P
DMEM High Glucose w/o L-Glutamine w/ 25mM Hepes w/o Sodium Pyruvate - 500ml L0100-500P
DMEM High Glucose w/o L-Glutamine w/ Sodium Pyruvate - 500ml L0106-500P
DMEM High Glucose w/o L-Glutamine w/o Sodium Pyruvate - 500ml L0101-500P
DMEM Low Glucose w/ L-Glutamine w/o Sodium Bicarbonate w/ Sodium Pyruvate - For 10L P0061-N10LP
DMEM Low Glucose w/ L-Glutamine w/o Sodium Bicarbonate w/ Sodium Pyruvate - For 1L P0061-N1LP
DMEM Low Glucose w/ L-Glutamine w/o Sodium Bicarbonate w/ Sodium Pyruvate - For 50L P0061-N50LP
DMEM Low Glucose w/ L-Glutamine w/o Sodium Bicarbonate w/ Sodium Pyruvate - For 5L P0061-N5LP
DMEM Low Glucose w/ L-Glutamine w/ Sodium Pyruvate - 500ml L0060-500P
DMEM Low Glucose w/ L-Glutamine w/ Sodium Pyruvate w/ 25 mM Hepes - 500ml L0065-500P
DMEM Low Glucose w/ Stable Glutamine w/ Sodium Pyruvate - 500ml L0066-500P
DMEM Low Glucose w/o L-Glutamine w/ Sodium Pyruvate - 500ml L0064-500P
MEM w/ Earle's Salts w/ L-Glutamine w/o NEAA w/o Sodium Bicarbonate - For 1L P0451-N1LP
RPMI 1640 w/o L-Glutamine w/o Sodium Bicarbonate w/o Phenol Red - For 1L P0871-N1LP
RPMI 1640 w/o L-Glutamine w/o Sodium Bicarbonate w/o Phenol Red - For 50L P0871-N50LP
RPMI 1640 w/o L-Glutamine w/o Sodium Bicarbonate w/o Phenol Red - For 10L P0871-N10LP
RPMI 1640 Dutch Modification w/o L-Glutamine w/ 1g/l Sodium Bicarbonate w/20mM Hepes - 500ml L0492-500P
RPMI 1640 w/ L- Glutamine w/o Sodium Bicarbonate w/ 25 mM Hepes w/o Phenol Red - For 10L P0876-N10LP
RPMI 1640 w/ L- Glutamine w/o Sodium Bicarbonate w/ 25 mM Hepes w/o Phenol Red - For 1L P0876-N1LP
RPMI 1640 w/ L- Glutamine w/o Sodium Bicarbonate w/ 25 mM Hepes w/o Phenol Red - For 50L P0876-N50LP
RPMI 1640 w/ L-Glutamine - 100ml L0500-100P
RPMI 1640 w/ L-Glutamine - 500ml L0500-500P
RPMI 1640 w/ L-Glutamine w/ 25 mM Hepes - 500ml L0495-500P
RPMI 1640 w/ L-Glutamine w/o Sodium Bicarbonate - For 100L P0860-N100LP
RPMI 1640 w/ L-Glutamine w/o Sodium Bicarbonate - For 10L P0860-N10LP
RPMI 1640 w/ L-Glutamine w/o Sodium Bicarbonate - For 1L P0860-N1LP
RPMI 1640 w/ L-Glutamine w/o Sodium Bicarbonate - For 50L P0860-N50LP
RPMI 1640 w/ L-Glutamine w/o Sodium Bicarbonate - For 5L P0860-N5LP
RPMI 1640 w/ L-Glutamine w/o Sodium Bicarbonate w/o Glucose - For 10L P0883-N10LP
RPMI 1640 w/ L-Glutamine w/o Sodium Bicarbonate w/o Glucose - For 1L P0883-N1LP
RPMI 1640 w/ L-Glutamine w/o Sodium Bicarbonate w/o Phenol Red - For 10L P0880-N10LP
RPMI 1640 w/ L-Glutamine w/o Sodium Bicarbonate w/o Phenol Red - For 1L P0880-N1LP
RPMI 1640 w/ L-Glutamine w/o Sodium Bicarbonate w/o Phenol Red - For 50L P0880-N50LP
RPMI 1640 w/ stable Glutamine - 100ml L0498-100P
RPMI 1640 w/ stable Glutamine - 500ml L0498-500P
RPMI 1640 w/ stable Glutamine w/ 25 mM Hepes - 500ml L0496-500P
RPMI 1640 w/o L-Glutamine - 100ml L0501-100P
RPMI 1640 w/o L-Glutamine - 500ml L0501-500P
RPMI 1640 w/o L-Glutamine w/ 25 mM Hepes - 500ml L0490-500P
RPMI 1640 w/o L-Glutamine w/o Folic Acid - 500ml L0503-500P
RPMI 1640 w/o L-Glutamine w/o Phenol Red - 500ml L0505-500P
RPMI 1640 w/o L-Glutamine w/o Sodium Bicarbonate - For 10L P0870-N10LP
RPMI 1640 w/o L-Glutamine w/o Sodium Bicarbonate - For 1L P0870-N1LP
RPMI 1640 w/o L-Glutamine w/o Sodium Bicarbonate - For 50L P0870-N50LP