Would you like to log in again?
Precision liquid handling and personalised medicine
Precision liquid handling and personalised medicine
Come take a look inside the Biotix Robotics Laboratory and see how we ensure the best for you!
Whether you call it “personalised” or “precision” medicine, there is no doubt that precision is essential when it comes to creating and executing medical programs designed for individual patients’ needs. The stakes are too high and the end results of contaminated samples or imprecise measuring are just too severe.
Liquid handling automation plays a key role in the workflow of personalised medicine. Because of the high quality standards needed, performance and quality are top considerations when selecting the right pipette tip.
For high-throughput automation, it is important that robotic tips meet stringent quality manufacturing standards. The QC process must ensure batch to batch conformity as well as meet exacting requirements for straightness, sealing, purity, accuracy, and precision.
In order to manufacture robotic tips that meet the strict requirements of personalised medicine, Biotix has built an in-house Robotic Laboratory for validation and release testing. Unlike other manufacturers that only perform dimensional inspection to engineering drawings, Biotix requires both dimensional and functional testing.
A combination of specialised fixtures and functional application tests are used, and are executed using the specific automated liquid handling system for which the tips have been designed. Experienced technicians measure coefficient of variance (CV), total indicated runout (TIR), insertion force and many other performance tests on the robots. If a product fails any test it is rejected, ensuring that only the highest quality tips reach the lab.
In order to get the most value from the precious samples associated with personalised medicine, Biotix ultra-straight tips target the center of microplate wells precisely, delivering exceptional accuracy and precision without risk of jamming. All Biotix robotic tips make use of the standard definitions bundled in the automation platforms’ software program, and they are BioReady™ certified to be free of RNase, DNase, and Endotoxin (pyrogens).
Robotic tips in the Biotix line are compatible with automated liquid handling platforms from these popular brands:
Agilent®
Beckman®
Tecan®
Caliper®
BioTek®
PerkinElmer/Packard®
Moleclar Devices®
Liquid handling automation plays a key role in personalised medicine making performance and quality top considerations when selecting pipette tips.
For more information on Biotix robotic tips contact your VWR representative or visit www.vwr.com/biotix.
Description Volume Pk Cat. No. Promo price Quantity
96-FORMAT ROBOTIC TIPS FOR AGILENT® PLATFORMS
Robotic tip for Agilent VPrep/Bravo, Clear, 96 tips/rack, Pre-sterile 250μl 10 racks/pack
5 packs/case
732-2460
Filtered robotic tip for Agilent VPrep/Bravo, Clear, 96 tips/rack, Pre-sterile 250μl 10 racks/pack
5 packs/case
732-2461
96-FORMAT ROBOTIC TIPS FOR BECKMAN® PLATFORMS
Robotic tip for Beckman FX, 96 tips/rack, Clear, Pre-sterile 20μl 10 racks/pack
5 packs/case
732-2764
Filtered robotic tip for Beckman FX, 96 tips/rack, Clear, Pre-sterile 20μl 10 racks/pack
5 packs/case
732-2765
Robotic tip for Beckman FX, 96 tips/rack, Clear, Pre-sterile 90μl 10 racks/pack
5 packs/case
732-2767
Filtered robotic tip for Beckman FX, 96 tips/rack, Clear, Pre-sterile 50μl 10 racks/pack
5 packs/case
732-2768
Robotic tip for Beckman FX, 96 tips/rack, Clear, Pre-sterile 220μl 10 racks/pack
5 packs/case
732-2770
Filtered robotic tip for Beckman FX, 96 tips/rack, Clear, Pre-sterile 125μl 10 racks/pack
5 packs/case
732-2771
Robotic tip for Beckman SPAN-8, Black, 96 tips/rack, Pre-sterile 20μl 10 racks/pack
5 packs/case
732-2456
Robotic filter tip for Beckman SPAN-8, Black, 96 tips/rack, Pre-sterile 20μl 10 racks/pack
5 packs/case
732-2559
Robotic tip for Beckman SPAN-8, Black, 96 tips/rack, Pre-sterile 250μl 10 racks/pack
5 packs/case
732-2458
Robotic filter tip for Beckman SPAN-8, Black, 96 tips/rack, Pre-sterile 250μl 10 racks/pack
5 packs/case
732-2560
96-FORMAT ROBOTIC TIPS FOR TECAN® PLATFORMS
Robotic tip for Tecan Temo/EVO, Clear, 96 tips/rack, Pre-sterile 50μl 10 racks/pack
5 packs/case
732-2426
Robotic tip for Tecan Temo/EVO, Clear, 96 tips/rack, Pre-sterile 200μl 10 racks/pack
5 packs/case
732-2428
Robotic Tip for Tecan Genesis, Hanging (LiHa) Liquid Sensing, Black, 96 tips/rack, Pre-sterile 20μl 24 racks/case 732-2430
Robotic Tip for Tecan Genesis, Hanging (LiHa) Liquid Sensing, Black, 96 tips/rack, Pre-sterile 200μl 24 racks/case 732-2432
Robotic Tip for Tecan Genesis, Hanging (LiHa) Liquid Sensing, Black, 96 tips/rack; Pre-sterile 1000μl 16 racks/case 732-2434
Robotic tip for Tecan Genesis, Hanging (LiHa) Clear, 96 tips/rack, Pre-sterile 20μl 24 racks/case 732-2436
Robotic tip for Tecan Genesis, Hanging (LiHa) Clear, 96 tips/rack, Pre-sterile 200μl 24 racks/case 732-2438
Robotic tip for Tecan Genesis, Hanging (LiHa) Clear, 96 tips/rack, Pre-sterile 1000μl 16 racks/case 732-2440
Robotic tip for MCA96, Clear, 8 nested trays/pack, Pre-sterile 50μl 8 trays/pack
10 packs/case
732-2442
Robotic tip for MCA96, Clear, 8 nested trays/pack, Pre-sterile 200μl 8 trays/pack
10 packs/case
732-2444
Robotic tip for MCA96, Clear, 96 tips/rack, Pre-sterile 50μl 8 trays/pack
10 packs/case
732-2446
Robotic tip for MCA96, Clear, 96 tips/rack, Pre-sterile 200μl 8 trays/pack
10 packs/case
732-2448
384-FORMAT ROBOTIC TIPS FOR AGILENT® PLATFORMS
Robotic tip for Agilent Velocity 11, 384-head, Clear, Pre-sterile 30μl 10 racks/pack
5 packs/case
736-0409
Filtered robotic tip for Agilent Velocity 11, 384-head, Clear, Pre-sterile 15μl 10 racks/pack
5 packs/case
736-0614
Robotic tip for Agilent Velocity 11, 384-head, Clear, Pre-sterile 70μl 10 racks/pack
5 packs/case
736-0413
Filtered robotic tip for Agilent Velocity 11, 384-head, Clear, Pre-sterile 70μl 10 racks/pack
5 packs/case
736-0615
384-FORMAT ROBOTIC TIPS FOR BECKMAN® PLATFORMS
Robotic tip for Beckman FX/NX, 384-head, Clear, Pre-sterile 30μl 10 racks/pack
5 packs/case
736-0410
Filtered robotic tip for Beckman FX/NX, 384-head, Clear, Pre-sterile 30μl 10 racks/pack
5 packs/case
736-0415
Robotic tip for Beckman FX/NX, 384-head, Clear, Pre-sterile 50μl 10 racks/pack
5 packs/case
736-0411
Filtered robotic tip for Beckman FX/NX, 384-head, Clear, Pre-sterile 45μl 10 racks/pack
5 packs/case
736-0412
384-FORMAT ROBOTIC TIPS FOR MOLECULAR DEVICES® PLATFORMS
Robotic tip for Molecular Devices, 384-head, Clear, Pre-sterile 30μl 10 racks/pack
5 packs/case
736-0414
Robotic tip for Molecular Devices, 384-head, Black, Pre-sterile 30μl 10 racks/pack
5 packs/case
732-2414