Would you like to log in again?
Biological buffers
Biological buffers
Practical tips for preparing buffer solutions
  • Buffer characteristics
  • Useful pH range
  • Preparing buffer solutions
  • Common buffer solutions
Many biochemical processes are markedly impaired by even small changes in the concentrations of free H + ions. It is therefore usually necessary to stabilise the H + concentration in vitro by adding a suitable buffer to the medium, without, however, affecting the functioning of the system under investigation.
A buffer keeps the pH value of a solution constant by taking up protons that are released during reactions, or by releasing protons when they are consumed by reactions.