Would you like to log in again?
ThawSTAR™ Automated Cell Thawing System
ThawSTAR™ Automated Cell Thawing System
Standardised thawing using breakthrough solid state technology
ThawSTAR automated sample thawing system addresses the last gap in the cell cryopreservation workflow: cell thawing. Replacing insufficient methods of thawing such as swirling samples in communal water baths and warming samples between hands - methods that are not standardized and potentially jeopardize the integrity and safety of the sample - ThawSTAR thawing system provides rapid, consistent and reproducible thawing and can be used in a biosafety cabinet
Check out this video, which walks you through the standardisation and workflow efficiency attainable with the ThawSTAR Transport and Thawing System.