Would you like to log in again?
Cleanroom Wipes
Sterile wipes clean aseptic cleanrooms quickly and easily withouth compromising the sterile nature of those environments. They are ideal for cleaning pharmaceutical aseptic fill areas and other sterile suites. "Ready to use" without time consuming and cost intensive treatment.
Dry cleanroom wipes are available for a variety of cleaning applications and in different materials (polyester, polyamide, nonwoven, microfibre, polypropylene).
Pre-wetted cleanroom wipes are effective and also save time because they are "ready-to-use" with the correct dosage / concentration of detergent. Spray flasks become unnecessary and the presence of volatiles is minimised.