Would you like to log in again?
Cleanroom Tapes
Tapes
Adhesive cleanroom tapes are suitable for colour marking, marking of surfaces, equipment, machinery, small parts and they are also suitable for barcode labels. They have a strong grip, but can be removed without residue. Very low generation of particles and outgassing, core is made of plastic. Available in different colours and different widths and lengths.
We recommend cuff stripes for an effective seal between glove and garment. Low-tack acrylic adhesive removes cleanly and leaves no adhesive residue or damage to garments. With heavy liner for easy separation, gamma irradiated.
Our recommendation:
Cuff stripes for an effective seal between glove and garment.