Would you like to log in again?
Cleanroom Sponges
Sponges
Sponges are ideal for the removal of strong and pasty contamination. They are available autoclavable and with material resistant to strong disinfectants.
For cleaning cleanroom curtains and softwalls, laminar flow benches and isolators. Also suitable as an intermediate layer for transport of critical products.
In a variety of sizes (including customised) and in different packaging units available.