Would you like to log in again?
Special Cleanroom Accessories
Special Accessories
In addition to the removal of contaminants and particles with wipes, other cleaning tasks require special accessories.
Examples include bench top protectors (with a low linting stabile surface, a higly absorbent middle layer and an impermeable bottom layer). The 3 layer construction allows a secure cytotoxic production and a save disposal. The integrated fluid barrier resists harsh chemicals. The slip-resistant polyethylene backing keeps mats in place. The mat lies flat and remains flat even after drying.      
Mitt wipes simplify the secure cleaning of hard to reach areas like carts, shelving or tubing in cleanrooms and controlled environments. For increased absorbency mitt wipes are available with foam interior. 