Would you like to log in again?
Permanent Cleanroom Mats
Permanent Mats Permanent mats are installed in cleanrooms to provide the highest level of cleanliness. The life of the mats strongly depends on the volume and degree of cleanliness of the foot and wheeled traffic.
Solvent-based cleaning materials should on no account be used on these mats: they will adversely affect both the function and the appearance. To avoid risk of slipping, it is also important that both the mats and the flooring should be completely dry.